Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 5 | 483 | ||
GenBank mRNA | AF181116.1 | - | 5 | 439 | ||
AK002241.1 | - | 5 | 484 | |||
AK002266.1 | - | 5 | 462 | |||
AK004013.1 | - | 5 | 439 | |||
AK005348.1 | - | 5 | 475 | |||
AK013391.1 | - | 5 | 484 | |||
AK014421.1 | - | 5 | 441 | |||
AK014435.1 | - | 5 | 713 | |||
AK135943.1 | - | 5 | 454 | |||
AK170514.1 | - | 5 | 500 | |||
BC024365.1 | - | 5 | 445 | 4988147 | ||
BC103767.1 | - | 5 | 486 | 30840199 |