Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018867.5 | NP_061355.3 | 14 | 3410 | ||
GenBank mRNA | AK019509.1 | - | 14 | 3409 |