Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019421.3 | NP_062294.3 | 5 | 759 | ||
GenBank mRNA | AK021187.1 | - | 5 | 734 | ||
AK078151.1 | - | 5 | 760 | |||
AK088189.1 | - | 5 | 760 | |||
AK131655.1 | - | 5 | 765 | |||
AK140204.1 | - | 5 | 746 | |||
BC026888.1 | - | 5 | 713 | 3991556 | ||
KJ650205.1 | - | 5 | 679 | |||
KJ663715.1 | - | 5 | 637 |