Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161428.1 | NP_001154900.1 | 7 | 870 | ||
NM_019449.2 | NP_062322.2 | 7 | 870 | |||
GenBank mRNA | AJ422144.1 | - | 1 | 12 | ||
AK030488.1 | - | 7 | 831 | |||
AK155556.1 | - | 7 | 828 | |||
AK162535.1 | - | 7 | 823 | |||
AK165060.1 | - | 7 | 830 | |||
BC018388.1 | - | 7 | 837 | 4209296 | ||
BC023731.1 | - | 1 | 295 | 5352340 |