Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019455.4 | NP_062328.3 | 6 | 876 | ||
GenBank mRNA | AK020246.1 | - | 6 | 877 | ||
AK039392.1 | - | 6 | 877 |