Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019519.2 | NP_062392.1 | 4, 5 | 758 | ||
GenBank mRNA | AF127656.1 | - | 5, 6 | 606 | ||
AF127657.1 | - | 4, 5 | 549 | |||
AF127658.1 | - | 4, 5 | 758 | |||
AF127659.1 | - | 4, 5 | 758 | |||
AF127660.1 | - | 5, 6 | 606 | |||
AF127661.1 | - | 4, 5 | 549 | |||
AF127662.1 | - | 5, 6 | 677 | |||
AK002625.1 | - | 5, 6 | 540 | |||
AK146917.1 | - | 5, 6 | 629 | |||
BC012214.1 | - | 5, 6 | 611 | 3967218 |