Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159616.1 | NP_001153088.1 | 5 | 542 | ||
NM_001159617.1 | NP_001153089.1 | 5 | 711 | |||
NM_001159618.1 | NP_001153090.1 | 5 | 621 | |||
NM_001159619.1 | NP_001153091.1 | 4 | 574 | |||
NM_019543.3 | NP_062416.1 | 5 | 557 | |||
GenBank mRNA | AF216306.1 | - | 5 | 544 | ||
AF216307.1 | - | 5 | 519 | |||
AF216308.1 | - | 4 | 464 | |||
AF216309.1 | - | 5 | 522 | |||
AK004169.1 | - | 5 | 521 | |||
BC038252.1 | - | 5 | 722 | 4913346 | ||
BC061116.1 | - | 5 | 495 | 6593482 | ||
BC096569.1 | - | 5 | 549 | 1429742 | ||
CB596599.1 | - | 4 | 578 |