Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019667.2 | NP_062641.1 | 4 | 329 | ||
GenBank mRNA | AB012611.1 | - | 4 | 344 | ||
AK004604.2 | - | 4 | 329 | |||
AK035789.1 | - | 382 | ||||
AK078244.1 | - | 4 | 361 | |||
AK159699.1 | - | 4 | 358 | |||
AK168729.1 | - | 4 | 361 | |||
BC013818.1 | - | 4 | 365 | 3671426 | ||
DQ141211.1 | - | 5 | 559 |