Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006841.5 | NP_006832.1 | 3 | 327 | ||
XM_006712954.2 | XP_006713017.1 | 3 | 361 | |||
GenBank mRNA | AF244548.1 | - | 3 | 286 | ||
AK299677.1 | - | 3 | 305 | |||
AK313461.1 | - | 3 | 301 | |||
BC042875.1 | - | 3 | 299 | 5174617 | ||
BP362985.1 | - | 3 | 327 | |||
BX537382.1 | - | 3 | 301 | |||
CR456970.1 | - | 2 | 173 | |||
U49082.1 | - | 3 | 297 |