Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019731.1 | NP_062705.1 | 5 | 627 | ||
GenBank mRNA | AF153451.1 | - | 5 | 627 | ||
AF288690.1 | - | 5 | 640 | |||
AK012805.1 | - | 5 | 663 | |||
AK017724.1 | - | 5 | 643 | |||
BC027277.1 | - | 5 | 645 | 3591383 |