Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285891.1 | NP_001272820.1 | 2, 3 | 367 | ||
NM_001285892.1 | NP_001272821.1 | 2, 3 | 367 | |||
NM_019748.3 | NP_062722.1 | 2, 3 | 367 | |||
GenBank mRNA | AB024303.1 | - | 2, 3 | 255 | ||
AK010313.1 | - | 2, 3 | 244 | |||
AK011772.1 | - | 2, 3 | 278 | |||
AK087556.1 | - | 2, 3 | 258 | |||
AK090012.1 | - | 2, 3 | 254 | |||
AK154139.1 | - | 2, 3 | 280 | |||
AK159672.1 | - | 2, 3 | 269 | |||
AK162789.1 | - | 2, 3 | 255 | |||
AK188153.1 | - | 1, 2 | 98 | |||
BB838763.1 | - | 2, 3 | 368 | |||
BC068164.1 | - | 2, 3 | 256 | 30359189 |