Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019748.3 | NP_062722.1 | 9 | 1605 | ||
GenBank mRNA | AB024303.1 | - | 9 | 1493 | ||
AK010313.1 | - | 9 | 1482 | |||
AK011783.1 | - | 6 | 952 | |||
AK087556.1 | - | 9 | 1496 | |||
AK090012.1 | - | 9 | 1492 | |||
AK159672.1 | - | 9 | 1507 | |||
BC068164.1 | - | 9 | 1494 | 30359189 |