Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168225.1 | NP_001161697.1 | 4 | 1776 | ||
NM_001168226.1 | NP_001161698.1 | 4 | 1709 | |||
NM_001168227.1 | NP_001161699.1 | 4 | 1661 | |||
NM_001168228.1 | NP_001161700.1 | 4 | 1660 | |||
NM_001168229.1 | NP_001161701.1 | 3 | 1632 | |||
NM_001168230.1 | NP_001161702.1 | 3 | 1570 | |||
NM_019768.4 | NP_062742.4 | 4 | 1783 | |||
GenBank mRNA | AB025049.1 | - | 2 | 1471 | ||
AK075946.1 | - | 4 | 1636 | |||
AK088849.1 | - | 1 | 1489 | |||
AK089963.1 | - | 4 | 1637 | |||
AK129038.1 | - | 4 | 1617 | |||
AK131944.1 | - | 4 | 1636 | |||
AK145686.1 | - | 4 | 1636 | |||
AK150401.1 | - | 4 | 1636 | |||
AK150670.1 | - | 4 | 1636 | |||
AK151570.1 | - | 4 | 1636 | |||
AK153146.1 | - | 4 | 1631 | |||
AK159145.1 | - | 4 | 1685 | |||
AK159179.1 | - | 4 | 1636 | |||
AK160012.1 | - | 4 | 1636 | |||
AK161610.1 | - | 4 | 1662 | |||
AK164433.1 | - | 4 | 1637 | |||
AK165985.1 | - | 4 | 1636 | |||
AK166637.1 | - | 4 | 1661 | |||
AK200823.1 | - | 1 | 90 | |||
BC075653.1 | - | 4 | 1783 | 30539330 | ||
BC088731.1 | - | 1 | 1417 | 5340043 |