Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019873.2 | NP_063926.1 | 2 | 248 | ||
GenBank mRNA | AK146018.1 | - | 2 | 318 | ||
BC046338.1 | - | 2 | 248 | 6395929 |