Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019964.1 | NP_064348.1 | 1 | 803 | ||
GenBank mRNA | AB028856.1 | - | 1 | 803 | ||
AK006026.1 | - | 1 | 787 | |||
BC049591.1 | - | 1 | 781 | 6773959 | ||
BC061112.1 | - | 1 | 786 | 6772873 |