Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brinp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019967.2 | NP_064351.2 | 4 | 856 | ||
GenBank mRNA | AB060589.1 | - | 4 | 856 | ||
AF202896.1 | - | 3 | 422 | |||
AK028321.1 | - | 4 | 771 | |||
AK045420.1 | - | 4 | 738 | |||
BC048789.1 | - | 4 | 824 | |||
BC079630.1 | - | 4 | 823 | 5694980 |