Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019968.2 | NP_064352.2 | 4 | 848 | ||
GenBank mRNA | AF205936.1 | - | 4 | 796 | ||
BC115498.2 | - | 4 | 789 | 40100921 |