Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177373.1 | NP_001170844.1 | 4 | 650 | ||
NM_019974.2 | NP_064358.1 | 3 | 616 | |||
GenBank mRNA | AB016226.1 | - | 4 | 500 | ||
AB016227.1 | - | 4 | 610 | |||
AK009360.1 | - | 4 | 651 | |||
AK009659.1 | - | 4 | 625 | |||
AK009720.1 | - | 4 | 651 | |||
BC115518.2 | - | 4 | 486 | |||
BC148310.2 | - | 3 | 325 |