Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019977.2 | NP_064361.2 | 3, 4 | 215 | ||
GenBank mRNA | AK002406.1 | - | 3, 4 | 229 | ||
AY738257.1 | - | 2, 3 | 185 | |||
BC013543.1 | - | 3, 4 | 215 | 4242159 |