Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006768.4 | NP_006759.3 | 7, 8 | 1181 | ||
XM_005253944.4 | XP_005254001.1 | 7, 8 | 1281 | |||
XM_011538789.2 | XP_011537091.1 | 6, 7 | 951 | |||
XM_017019992.1 | XP_016875481.1 | 6, 7 | 989 | |||
XM_017019993.1 | XP_016875482.1 | 6, 7 | 972 | |||
XM_017019994.1 | XP_016875483.1 | 5, 6 | 847 | |||
XM_017019995.1 | XP_016875484.1 | 5, 6 | 612 | |||
GenBank mRNA | AB208878.1 | - | 7, 8 | 1136 | ||
AF035620.1 | - | 7, 8 | 1116 | |||
AK299326.1 | - | 4, 5 | 487 | |||
AK308279.1 | - | 7, 8 | 1028 | |||
AY332222.1 | - | 7, 8 | 1116 | |||
BC136698.1 | - | 7, 8 | 1116 | 9020688 | ||
BC136699.1 | - | 7, 8 | 1116 | 9020689 |