Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020333.2 | NP_065066.2 | 17 | 2469 | ||
GenBank mRNA | AF332063.1 | - | 17 | 2139 | ||
AF332064.1 | - | 17 | 2139 | |||
AK122460.1 | - | 17 | 2203 | |||
AK147262.1 | - | 17 | 2471 | |||
BC054808.1 | - | 17 | 2135 | 6838880 | ||
KJ535320.1 | - | 17 | 2160 | |||
KJ535321.1 | - | 17 | 2160 | |||
KJ535322.1 | - | 17 | 2316 |