Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000956.3 | NP_000947.2 | 2 | 1337 | ||
GenBank mRNA | BC093927.1 | - | 2 | 1139 | 7939772 | |
BC093929.1 | - | 2 | 1139 | 7939774 | ||
BF029797.1 | - | 177 | ||||
CF132756.1 | - | 1 | 109 | |||
U19487.1 | - | 2 | 1242 |
Set Membership: |
G-protein coupled receptor |