Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc46a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021053.4 | NP_066395.3 | 1 | 1192 | ||
GenBank mRNA | AF148145.1 | - | 1 | 1164 | ||
AK014784.1 | - | 1 | 1173 | |||
AK017363.1 | - | 1 | 1192 | |||
AK040063.1 | - | 1193 | ||||
BC140962.1 | - | 1 | 1125 | 9055574 |