Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113356.1 | NP_001106827.1 | 794 | |||
GenBank mRNA | AF459018.1 | - | 794 |
Gene Symbol: C1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023143.3 | NP_075632.3 | 5, 6 | 945 | ||
GenBank mRNA | AF148216.1 | - | 5, 6 | 933 | ||
AK138817.1 | - | 5, 6 | 844 | |||
BC004637.1 | - | 5, 6 | 887 | 3586055 | ||
BC093528.1 | - | 5, 6 | 1161 | 5135606 | ||
BC172078.1 | - | 5, 6 | 870 | 9054364 |