Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026203.3 | NP_080479.2 | 4 | 977 | ||
GenBank mRNA | AK005991.1 | - | 4 | 919 | ||
AK036336.1 | - | 4 | 867 | |||
AY133241.1 | - | 4 | 870 | |||
BC055400.1 | - | 3 | 699 | 6467369 | ||
BC065146.1 | - | 4 | 873 |