Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lurap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026821.5 | NP_081097.2 | 2 | 981 | ||
GenBank mRNA | AK003950.1 | - | 2 | 290 | ||
BC021501.1 | - | 2 | 865 | 5346437 | ||
BC030404.1 | - | 2 | 982 | 5354204 |