Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133791.4 | NP_598552.2 | 7 | 1113 | ||
GenBank mRNA | AB045323.1 | - | 7 | 1136 | ||
AK032989.1 | - | 3 | 179 | |||
AK079375.1 | - | 7 | 948 | |||
AK086956.1 | - | 7 | 943 | |||
AK147648.1 | - | 1107 | ||||
AK147944.1 | - | 7 | 1082 | |||
AK186508.1 | - | 5 | 359 | |||
AK211421.1 | - | 2 | 61 | |||
BC067050.1 | - | 7 | 1113 | 30537299 |