Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133791.4 | NP_598552.2 | 8 | 1256 | ||
GenBank mRNA | AB045323.1 | - | 8 | 1279 | ||
AK032989.1 | - | 4 | 322 | |||
AK079375.1 | - | 8 | 1091 | |||
AK086956.1 | - | 8 | 1086 | |||
AK147648.1 | - | 1250 | ||||
AK147944.1 | - | 8 | 1225 | |||
AK209117.1 | - | 2 | 142 | |||
AK211421.1 | - | 3 | 204 | |||
BC067050.1 | - | 8 | 1256 | 30537299 |