Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138592.4 | NP_613058.1 | 5 | 652 | ||
GenBank mRNA | AK010443.1 | - | 5 | 662 | ||
AK018595.1 | - | 5 | 653 | |||
AK033105.1 | - | 5 | 649 | |||
AK045062.1 | - | 5 | 649 | |||
AK088354.1 | - | 5 | 664 | |||
AK165625.1 | - | 5 | 633 | |||
AK167668.1 | - | 5 | 640 | |||
AK168003.1 | - | 5 | 659 | |||
AK206422.1 | - | 1 | 61 | |||
BC026983.1 | - | 5 | 627 | 5365862 |