Search Thermo Fisher Scientific
Gene Symbol: COPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006833.4 | NP_006824.2 | 3, 4 | 363 | ||
GenBank mRNA | AK312024.1 | - | 3, 4 | 354 | ||
BC002520.2 | - | 3, 4 | 337 | 3140042 | ||
BI113801.1 | - | 3, 4 | 364 | |||
CB990467.1 | - | 3, 4 | 387 | |||
U70735.1 | - | 3, 4 | 285 |