Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 7 | 2595 | ||
GenBank mRNA | AK005457.1 | - | 1 | 532 | ||
AK045714.1 | - | 6 | 3095 | |||
AY453793.1 | - | 7 | 2597 | |||
BC030919.1 | - | 2 | 2688 | |||
BC042657.1 | - | 4 | 5765 |