Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145528.3 | NP_663503.1 | 13 | 1416 | ||
GenBank mRNA | AK122339.1 | - | 9 | 3778 | ||
AK149672.1 | - | 13 | 1375 | |||
AK154470.1 | - | 13 | 1404 | |||
AK163962.1 | - | 13 | 1370 | |||
AK171206.1 | - | 13 | 1402 | |||
AK171440.1 | - | 13 | 1393 | |||
AK181445.1 | - | 3 | 152 | |||
BC016669.1 | - | 13 | 1400 | 2648797 | ||
BC023673.1 | - | 13 | 1366 | 5343236 | ||
BC023712.1 | - | 13 | 1389 | 5346303 | ||
BC033419.1 | - | 13 | 1367 | 5359944 |