- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038608.2 | NP_001033697.2 | 4 | 1261 | ||
NM_001042408.1 | NP_001035867.1 | 4 | 1237 | |||
NM_178604.3 | NP_848719.2 | 4 | 1373 | |||
GenBank mRNA | AK010599.1 | - | 4 | 1261 | ||
AK011717.1 | - | 4 | 1325 | |||
AK012775.1 | - | 1 | 187 | |||
AK180869.1 | - | 1 | 154 | |||
AK186245.1 | - | 1 | 154 | |||
AK205305.1 | - | 1 | 154 | |||
BC003876.1 | - | 4 | 1178 | |||
BC014770.1 | - | 4 | 1237 | |||
BC025053.1 | - | 4 | 1203 | |||
BC029327.1 | - | 4 | 1220 |