Gene Symbol: Map4k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008279.2 | NP_032305.2 | 20, 21 | 1565 | ||
GenBank mRNA | AK143215.1 | - | 21, 22 | 1649 | ||
AK146273.1 | - | 20, 21 | 1549 | |||
BC005433.1 | - | 20, 21 | 1558 | 3494470 | ||
BC024832.1 | - | 7, 8 | 422 | |||
Y09010.1 | - | 20, 21 | 1601 |