Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 12 | 1287 | ||
GenBank mRNA | AK008141.1 | - | 12 | 1290 | ||
AK011002.1 | - | 12 | 1288 | |||
AK150067.1 | - | 12 | 1288 | |||
AK159379.1 | - | 12 | 1286 | |||
AK166255.1 | - | 12 | 1289 | |||
AK166507.1 | - | 12 | 1288 | |||
AK167209.1 | - | 12 | 1288 | |||
AK167442.1 | - | 12 | 1291 | |||
AK168710.1 | - | 12 | 1270 | |||
AK176968.1 | - | 3 | 257 | |||
BC007467.1 | - | 11 | 1226 | 3497714 |