Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 4 | 1434 | ||
GenBank mRNA | AK096906.1 | - | 2 | 459 | ||
AK291996.1 | - | 4 | 1434 | |||
AK300649.1 | - | 4 | 1232 | |||
AY322545.1 | - | 4 | 839 | |||
BC096842.1 | - | 4 | 845 | 6971906 | ||
FJ200479.1 | - | 5 | 731 | |||
M57414.1 | - | 4 | 839 |
Set Membership: |
G-protein coupled receptor |