Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009320.4 | NP_033346.2 | 15 | 3896 | ||
GenBank mRNA | L03292.2 | - | 15 | 3871 |