Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142918.1 | NP_001136390.1 | 1 | 656 | ||
NM_001142919.1 | NP_001136391.1 | 1 | 656 | |||
NM_001142920.1 | NP_001136392.1 | 1 | 656 | |||
NM_001142921.1 | NP_001136393.1 | 1 | 656 | |||
NM_001142922.1 | NP_001136394.1 | 1 | 656 | |||
NM_001142923.1 | NP_001136395.1 | 1 | 656 | |||
NM_009333.3 | NP_033359.2 | 1 | 656 | |||
GenBank mRNA | AF107298.1 | - | 1 | 400 | ||
AF363724.1 | - | 1 | 212 | |||
AF363725.1 | - | 1 | 214 | |||
AF363726.1 | - | 1 | 218 | |||
AJ223070.1 | - | 1 | 134 | |||
AK035059.1 | - | 1 | 484 | |||
AK038720.1 | - | 1 | 102 | |||
AK084488.1 | - | 1 | 197 | |||
AK132465.1 | - | 1 | 585 | |||
AY072035.1 | - | 1 | 197 | |||
BC009807.1 | - | 1 | 161 | |||
BC011397.1 | - | 1 | 635 | |||
BC052022.1 | - | 1 | 235 | 5702449 |