Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011550.1 | - | 8 | 956 | ||
AK076365.1 | - | 6 | 1872 | |||
AK133474.1 | - | 8 | 924 | |||
AK139088.1 | - | 8 | 1162 | |||
AK144793.1 | - | 8 | 1140 | |||
AK155268.1 | - | 6 | 1901 | |||
AK193704.1 | - | 1 | 26 | |||
BC015077.1 | - | 8 | 838 | |||
BC145595.1 | - | 7 | 930 | |||
BC145596.1 | - | 8 | 845 |