Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009344.3 | NP_033370.2 | 2 | 1592 | ||
GenBank mRNA | AK148063.1 | - | 2 | 1328 | ||
AK158890.1 | - | 2 | 1476 | |||
BC010295.1 | - | 2 | 1155 | 3153799 | ||
U44088.1 | - | 2 | 1592 |