Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291765.1 | NP_001278694.1 | 4 | 276 | ||
NM_001291766.1 | NP_001278695.1 | 3 | 270 | |||
NM_001291768.1 | NP_001278697.1 | 3 | 209 | |||
NM_009361.3 | NP_033387.1 | 4 | 337 | |||
GenBank mRNA | AK013180.1 | - | 3 | 239 | ||
AK019997.1 | - | 4 | 277 | |||
AK030820.1 | - | 4 | 338 | |||
AK154037.1 | - | 3 | 177 | |||
BC058633.1 | - | 4 | 320 | |||
BC132567.1 | - | 3 | 182 | 40130844 | ||
BC132569.1 | - | 3 | 182 | 40130846 | ||
BC145344.1 | - | 3 | 182 | 9053900 | ||
BY089512.1 | - | 3 | 186 | |||
X72310.1 | - | 3 | 211 |