Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291765.1 | NP_001278694.1 | 6, 7 | 595 | ||
NM_001291766.1 | NP_001278695.1 | 5, 6 | 589 | |||
NM_001291768.1 | NP_001278697.1 | 5, 6 | 528 | |||
NM_009361.3 | NP_033387.1 | 6, 7 | 656 | |||
GenBank mRNA | AK013180.1 | - | 5, 6 | 558 | ||
AK019997.1 | - | 6, 7 | 596 | |||
AK030820.1 | - | 6, 7 | 657 | |||
AK154037.1 | - | 5, 6 | 496 | |||
BC132567.1 | - | 5, 6 | 501 | 40130844 | ||
BC132569.1 | - | 5, 6 | 501 | 40130846 | ||
BC145344.1 | - | 5, 6 | 501 | 9053900 | ||
X72310.1 | - | 5, 6 | 530 |