Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009367.3 | NP_033393.2 | 7 | 2957 | ||
GenBank mRNA | AK029306.1 | - | 5 | 1168 | ||
AK052377.1 | - | 1 | 403 | |||
AK084760.1 | - | 1 | 234 | |||
AK159690.1 | - | 7 | 1520 | |||
AK163286.1 | - | 1 | 403 | |||
BC011055.1 | - | 5 | 1318 | 3585774 | ||
X57413.1 | - | 7 | 2956 |