Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Th | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009377.1 | NP_033403.1 | 7 | 835 | ||
GenBank mRNA | AK139568.1 | - | 1052 | |||
M69200.1 | - | 7 | 835 |