Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
TNNT3
Gene Name:
troponin T3, fast skeletal type
Gene Aliases:
TNTF
Chromosome Location:
Chr.11: 1919551 - 1938706 on Build GRCh38
UniGene ID:
Hs.73454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18674
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNNT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042780.2
NP_001036245.1
10
519
NM_001042781.2
NP_001036246.1
11
537
NM_001042782.2
NP_001036247.1
10
519
NM_001297646.1
NP_001284575.1
11
777
NM_006757.3
NP_006748.1
11
543
XM_006718288.3
XP_006718351.1
6
570
XM_006718290.3
XP_006718353.1
13
482
XM_006718293.2
XP_006718356.1
12
559
XM_006718294.3
XP_006718357.1
12
559
XM_006718296.3
XP_006718359.1
11
446
XM_006718299.2
XP_006718362.1
10
382
XM_006718300.3
XP_006718363.1
9
410
XM_006718302.3
XP_006718365.1
8
392
XM_011520343.2
XP_011518645.1
13
613
XM_017018205.1
XP_016873694.1
11
537
XM_017018206.1
XP_016873695.1
13
405
XM_017018207.1
XP_016873696.1
12
393
XM_017018208.1
XP_016873697.1
11
360
XM_017018209.1
XP_016873698.1
11
369
XM_017018210.1
XP_016873699.1
10
334
XM_017018211.1
XP_016873700.1
10
353
XM_017018212.1
XP_016873701.1
9
325
XM_017018213.1
XP_016873702.1
8
310
XM_017018214.1
XP_016873703.1
8
309
XM_017018215.1
XP_016873704.1
8
301
XM_017018216.1
XP_016873705.1
12
464
XM_017018217.1
XP_016873706.1
11
443
XM_017018218.1
XP_016873707.1
10
431
XM_017018219.1
XP_016873708.1
8
392
GenBank mRNA
AK056968.1
-
10
598
BC050446.1
-
10
375
BC117327.1
-
9
403
40125878
BC143537.1
-
9
403
9052047
BC171727.1
-
10
421
9052046
BC171728.1
-
6
356
BT019997.1
-
10
325
BX640689.1
-
11
401
CR541927.1
-
10
325
DA897990.1
-
11
537
DA899275.1
-
9
428
DQ778624.1
-
6
307
M21984.1
-
10
343
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
regulation of striated muscle contraction
muscle filament sliding
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0