Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009390.2 | NP_033416.2 | 4 | 987 | ||
GenBank mRNA | AK080919.1 | - | 4 | 1003 | ||
U34042.1 | - | 4 | 987 |