Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009419.3 | NP_033445.2 | 5 | 1284 | ||
GenBank mRNA | AF049890.1 | - | 4 | 1230 | ||
AK036436.1 | - | 4 | 1213 | |||
AK075672.1 | - | 5 | 1284 | |||
AK149502.1 | - | 5 | 1249 | |||
AK150082.1 | - | 5 | 1244 | |||
AK163461.1 | - | 5 | 1248 | |||
AK190004.1 | - | 1 | 39 | |||
AK204832.1 | - | 1 | 39 | |||
BC003721.1 | - | 5 | 1275 | 3494892 |