Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009425.2 | NP_033451.1 | 5 | 527 | ||
GenBank mRNA | AK142960.1 | - | 5 | 529 | ||
AK144931.1 | - | 544 | ||||
AK153581.1 | - | 5 | 577 | |||
AK157633.1 | - | 528 | ||||
BC011469.1 | - | 5 | 493 | |||
BC138807.1 | - | 5 | 501 | 8861829 | ||
BC138808.1 | - | 5 | 501 | 8861830 | ||
U37522.1 | - | 5 | 503 |