Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169576.1 | NP_001163047.1 | 5 | 910 | ||
NM_001169577.1 | NP_001163048.1 | 5 | 486 | |||
NM_009459.3 | NP_033485.1 | 7 | 1003 | |||
GenBank mRNA | AK131688.1 | - | 7 | 981 | ||
AK161414.1 | - | 5 | 894 | |||
BC008517.1 | - | 7 | 809 | 3600233 | ||
U19854.1 | - | 7 | 732 |