Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289612.1 | NP_001276541.1 | 6 | 1029 | ||
NM_001289765.1 | NP_001276694.1 | 13 | 1976 | |||
NM_001289766.1 | NP_001276695.1 | 13 | 1941 | |||
NM_009539.3 | NP_033565.2 | 13 | 1933 | |||
GenBank mRNA | AB083210.1 | - | 6 | 1032 | ||
AB084383.1 | - | 6 | 871 | |||
AK039883.1 | - | 13 | 1870 | |||
AK089056.1 | - | 13 | 1905 | |||
BC029727.1 | - | 13 | 1891 | 4925739 | ||
U04379.1 | - | 13 | 1887 | |||
U77667.1 | - | 12 | 1779 |